Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-azido-7-hydroxychromen-2-one | 817638-68-9 | MFCD12924840 | 98.0% | 203.15 | C9H5N3O3 | 25 MG
SDP

Supplier:  Medchemexpress LLC HYW54097825MG

Encompass_Preferred

3-Azido-7-hydroxycoumarin is a fluorogenic click-chemistry reagent containing an azide group that undergoes Cu-catalyzed azide-alkyne cycloaddition to label biomolecules; the cycloaddition yields a fluorescent signal used for detection and imaging in biochemical assays. Supplied as a solid for laboratory use.

  • Fluorogenic probe for click chemistry.
  • High purity (98.0%).
  • Molecular weight 203.15 g/mol.
  • Chemical formula C9H5N3O3.
  • Appearance: solid, yellow to brown.
  • Storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-84188


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.