Learn More
Medchemexpress LLC 3-azido-7-hydroxycoumarin | 817638-68-9 | MFCD12924840 | >95.0% | 203.15 g/mol | C9H5N3O3 | 5 MG

Supplier: Medchemexpress LLC HYW5409785MG
3-Azido-7-hydroxycoumarin is a fluorogenic click chemistry reagent containing an azide functional group. It becomes fluorescent after copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC), enabling sensitive bioorthogonal labeling and probe development. Chemical identifiers: C9H5N3O3; MW 203.15 g/mol; CAS 817638-68-9.
- Fluorogenic upon copper(I)-catalyzed azide-alkyne cycloaddition.
- Excitation maximum 404 nm and emission maximum 477 nm after click reaction.
- Azide reactive handle for bioorthogonal labeling.
- Suitable for probe development and imaging applications.
- Molecular weight 203.15 g/mol; formula C9H5N3O3.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.