Learn More
Medchemexpress LLC 3-azido-7-hydroxycoumarin | 817638-68-9 | MFCD12924840 | 98.0% | 203.15 | C9H5N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW54097810MG
Encompass_Preferred
3-Azido-7-hydroxycoumarin is a coumarin-based fluorogenic reagent containing an azide functional group used for bioorthogonal labeling via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is intended for research use in fluorescence-based assays and imaging, supplied as a solid and provided in multiple pack sizes for laboratory applications.
  • Fluorogenic coumarin scaffold for sensitive detection.
  • Reactive azide group for CuAAC bioorthogonal labeling.
  • Suitable for fluorescence assays and cellular imaging.
  • Stable as a powder with recommended storage at -20°C.
  • Available in small lab-scale quantities for research use.
Catalog No. 50-002-84089