Learn More
Apexbio Technology LLC 3-AZIDOPROPYLAMINE-250MG
SDP
Supplier:  Apexbio Technology LLC BA2384250
Encompass_Preferred
3-Azidopropylamine (CAS No 88192-19-2) is a compound of interest in biomedical research due to its potential applications in drug discovery and development Originating from synthetic organic chemistry this molecule is studied for its ability to interact with biological targets through azide-functional group-mediated reactions which can facilitate bioconjugation and labeling Its mechanism involves pathways where azide groups participate in click chemistry making it valuable in modifying biomolecules Common in vitro activities of 3-azidopropylamine occur in nanomolar to low micromolar ranges demonstrating its efficacy in cell assays It is often utilized in cell models and animal studies for drug screening purposes Experimental concentrations typically depend on the specific research design allowing flexibility in application for various biomedical investigations
Catalog No. 50-005-68099