Learn More
Medchemexpress LLC 3-Azidopropylamine | 88192-19-2 | MFCD11046568 | C3H8N4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY151862
Encompass_Preferred
3-Azidopropylamine is a click chemistry reagent containing an azide group. It is utilized for the complexation and transfection of plasmid DNA by reacting with the starch sugar of potato starch. This reagent facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and can also undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Reacts with starch sugar for complexation and transfection of plasmid DNA
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-75948