Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(benzyloxy)oxan-4-one | 1351450-56-0 | 98.2% | 206.24 g/mol | C12H14O3 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0598615G

Encompass_Preferred

3-(benzyloxy)oxan-4-one is a chemical intermediate used in the synthesis of active pharmaceutical compounds and as a reference compound in medicinal chemistry research. It is supplied in small research-scale pack sizes and is typically provided at high purity for building-block applications.

  • Used as a drug intermediate and reference compound.
  • CAS number 1351450-56-0.
  • Molecular formula C12H14O3.
  • Molecular weight 206.24 g/mol.
  • Purity 98.2%.
  • Available in small pack sizes, including 5 G.

Catalog No. 50-002-10370


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.