Learn More
Medchemexpress LLC 3α-bis-(4-fluorophenyl) methoxytropane hydrochloride | 202646-03-5 | 100.0% | 379.87 | C21H24ClF2NO | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101315100MG
Encompass_Preferred
AHN 1-055 hydrochloride is a research-grade dopamine uptake inhibitor that binds with high affinity to the dopamine transporter (DAT). Supplied as the hydrochloride salt, it is intended for pharmacological and neuroscience studies requiring selective DAT inhibition.
  • High affinity DAT inhibitor; IC50 = 71 nM.
  • Supplied as hydrochloride salt for improved handling and stability.
  • Reported purity 99.96%.
  • Molecular formula C21H24ClF2NO; molecular weight 379.87.
  • Available as solid sizes and as DMSO solution for assay use.
  • Manufacturer-provided COA and SDS documentation available.
Catalog No. 50-002-11127