Learn More
Medchemexpress LLC Methyl (1R)-3-oxocyclopentane-1-carboxylate | 132076-27-8 | MFCD01320815 | 142.15 g/mol | C7H10O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW057459100MG
Encompass_Preferred
(R)-3-Oxo-cyclopentanecarboxylic acid methyl ester is a chiral synthetic intermediate used in medicinal chemistry and pharmaceutical synthesis. It provides the R-enantiomer of methyl 3-oxocyclopentane-1-carboxylate and is commonly used in stereospecific syntheses and route development.
  • Chiral R-enantiomer useful for stereospecific synthesis.
  • Molecular formula C7H10O3 and molecular weight 142.15 g/mol.
  • Commonly used as a synthetic intermediate in pharmaceutical research.
  • Available in small-scale packages for route development and optimization.
  • Documentation available for handling and characterization.
Catalog No. 50-002-10004