Learn More
Medchemexpress LLC Tert-butyl 9-formyl-3-azaspiro[5.5]undecane-3-carboxylate | 1416176-14-1 | 98.0% | 281.39 g/mol | C16H27NO3 | 50 MG

Supplier: Medchemexpress LLC HYW05626750MG
3-Boc-3-azaspiro[5.5]undecane-9-carbaldehyde is a Boc-protected spirocyclic aldehyde used as a building block and PROTAC linker for medicinal chemistry and conjugation chemistry. It provides a Boc-protected tertiary amine in a spiro scaffold and an aldehyde functional group for further derivatization.
- Boc-protected tertiary amine in a spirocyclic scaffold.
- Aldehyde functional group suitable for conjugation and linker formation.
- CAS number 1416176-14-1.
- Chemical formula C16H27NO3; molecular weight 281.39 g/mol.
- Purity 98.0% (NMR) for batch 439849; appearance colorless to off-white (solid-liquid mixture).
- Storage: store at -20°C under nitrogen; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.