Learn More
Medchemexpress LLC 3-bromo-1-(3-chloro-2-pyridinyl)-N-[4-cyano-2-methyl-6-(methylcarbamoyl)phenyl]pyrazole-5-carboxamid | 736994-63-1 | MFCD28053506 | 99.4% | 473.71 g/mol | C19H14BrClN6O2 | 5 MG

Supplier: Medchemexpress LLC HY12779R5MG
Cyantraniliprole is the analytical standard of the insecticidal active ingredient cyantraniliprole, provided as a research-grade reference material for analytical testing and method development. The compound acts as a ryanodine receptor agonist with insecticidal and antioxidant activity, and is supplied in small quantities for laboratory use.
- Purity 99.4%
- Molecular formula C19H14BrClN6O2
- Molecular weight 473.71 g/mol
- Intended for research and analytical applications
- Available in small quantities suitable for analytical standards
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.