Learn More
Medchemexpress LLC 3-bromo-1H-pyrrole-2,5-dione | 98026-79-0 | ≥97.0% | 175.97 | C4H2BrNO2 | 100 MG

Supplier: Medchemexpress LLC HYW076794100MG
3-Bromo-1H-pyrrole-2,5-dione is a 3-substituted pyrrole-2,5-dione compound reported to have antibacterial activity against several pathogenic strains. It is provided as a solid for research use and is available in multiple package sizes.
- Antibacterial activity against S. aureus, E. coli, and P. aeruginosa reported.
- Cas number 98026-79-0.
- Molecular formula C4H2BrNO2.
- Molecular weight 175.97.
- Purity ≥97.0%.
- Available in 100 mg to 10 g pack sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.