Learn More
Medchemexpress LLC 3-Bromo-2-methylaniline | 55289-36-6 | 99.8% | C7H8BrN | 10 G
SDP
Supplier:  Medchemexpress LLC HY5009710G
Encompass_Preferred
3-Bromo-2-methylaniline is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Prepare 4-bromo-1H-indazole
  • Prepare 3,4-dibromo-2-methylaniline
  • Prepare derivatives of Troger's base
Catalog No. 50-003-57925