Learn More
Medchemexpress LLC 3-bromo-2-methylpropanoic acid | 56970-78-6 | MFCD01310530 | ≥95.0% | 167.00 g·mol-1 | C4H7BrO2 | 1 G

Supplier: Medchemexpress LLC HYW0153321G
3-Bromo-2-methylpropanoic acid is an identified impurity of captopril provided for research and analytical use. Supplied as a solid and as a DMSO solution, it is used for impurity profiling, analytical method development, and as a synthetic intermediate for research purposes.
- Chemical name 3-bromo-2-methylpropanoic acid; CAS 56970-78-6.
- Purity ≥95.0%.
- Available as solid and 10 mM DMSO solution.
- Suitable for analytical reference standards and impurity profiling.
- Useful as a building block or intermediate in organic synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.