Learn More
Medchemexpress LLC 3-Bromo-5-chloropyridine | 73583-39-8 | 192.44 | 25 G

Supplier: Medchemexpress LLC 25GHYW003165
3-Bromo-5-chloropyridine is a widely used heterocyclic compound that can be used to synthesize various compounds. It serves as an intermediate in the production of crop protectants within the agricultural chemicals industry and can also participate in cross-coupling reactions.
- Appearance: Off-white to light brown solid
- Consistent with structure based on ¹H NMR spectrum
- Consistent with structure based on MS
- Purity (HPLC): 99.87%
- Water (KF): 0.09%
- Target: Biochemical assay reagents
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.