Learn More
eMolecules​ 1146615-86-2 | 3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine | Combi-Blocks | MFCD12827771 | 265.033 | C8H4BrF3N2 | 95.000 | FC(F)(F)c1ccc2ncc(Br)n2c1 | 5g | 401039363
Supplier:  eMolecules​ 401039363
Encompass_Preferred
3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine | Combi-Blocks | 1146615-86-2 | MFCD12827771 | 265.033 | C8H4BrF3N2 | 95.000 | FC(F)(F)c1ccc2ncc(Br)n2c1 | 5g | 401039363
Catalog No. NC3727479