Learn More
Medchemexpress LLC HY-101175 10mg Medchemexpress, 3-Bromo-7-nitroindazole CAS:74209-34-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10117510MG
Encompass_Preferred
Medchemexpress, HY-101175 10mg 3-Bromo-7-nitroindazole CAS:74209-34-0 3-Bromo-7-nitroindazole is a more potent and selective inhibitor of neuronal nitric oxide synthase (nNOS) than eNOS or inducible nitric oxide synthase (iNOS). 3-Bromo-7-nitroindazole affects the intercellular messenger nitric oxide (NO) synthesis throughout the body and brain [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2057