Learn More
Cayman Chemical 3-bromo-7-NItroIndazole 250mg
SDP
Supplier:  Cayman Chemical 81345250 MG
Encompass_Preferred
3-Bromo-7-nitroindazole is a more potent inhibitor of nNOS than 7-nitroindazole in vitro, but unlike 7-nitroindazole, it is also potent against iNOS. The IC50 values for inhibition of rat nNOS, bovine eNOS, and rat iNOS are 0.17, 0.86, and 0.29 µM, respectively.{1339}
Catalog No. 50-289-4600