Learn More
Apexbio Technology LLC 3-Bromo-7-nitroindazole 74209-34-0 10mg
SDP
Supplier:  Apexbio Technology LLC B642610
Encompass_Preferred
3-Bromo-7-nitroindazole is an indazole derivative structurally defined by a bromine and a nitro group substituted on the indazole scaffold It acts predominantly as a selective inhibitor of neuronal nitric oxide synthase (nNOS) thereby modulating the synthesis of nitric oxide in neural tissues Through nNOS inhibition 3-Bromo-7-nitroindazole is employed in biomedical research to investigate the physiological and pathological roles of nitric oxide particularly in studies related to neurodegeneration ischemia and inflammatory neural processes
Catalog No. 50-000-92223