Learn More
Medchemexpress LLC 3-bromo-7-nitroindazole | 74209-34-0 | MFCD00159910 | 98.1% | 242.03 g/mol | C7H4BrN3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10117510MM 1ML
Encompass_Preferred
3-bromo-7-nitroindazole is a selective neuronal nitric oxide synthase (nNOS) inhibitor provided as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for biochemical and cellular studies to modulate nitric oxide production. The compound has formula C7H4BrN3O2, molecular weight 242.03 g/mol, and reported purity of approximately 98%.
  • Selective neuronal nitric oxide synthase inhibitor.
  • Provided as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity for reliable experimental results.
  • Suitable for biochemical and cellular assays involving nitric oxide signaling.
  • Soluble in common organic solvents and supplied as a light yellow solid in dry form.
Catalog No. 50-002-12880