Learn More
Medchemexpress LLC 3-bromo-l-tyrosine | 38739-13-8 | MFCD09836116 | 98.7% | 260.09 g·mol⁻¹ | C9H10BrNO3 | 10 G

Supplier: Medchemexpress LLC HYW01838610G
3-bromo-l-tyrosine is a brominated derivative of the amino acid l-tyrosine used as a research reagent and building block in biochemical and medicinal chemistry studies. It is supplied as a solid for laboratory research use only and is characterized by high purity, a defined molecular weight, and specified storage recommendations.
- High purity: 98.7%.
- Molecular formula: C9H10BrNO3.
- Molecular weight: 260.09 g·mol⁻¹.
- Physical form: off-white to light brown solid.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Documentation available: datasheet, certificate of analysis, and safety data sheet.
- Available in multiple pack sizes suitable for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.