Learn More
Medchemexpress LLC 3-Bromocarbazole | 1592-95-6 | 99.7% | 246.10 | 25 G

Supplier: Medchemexpress LLC 25GHY33798
3-Bromocarbazole is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
- Cas no.: 1592-95-6
- Molecular weight: 246.10
- Purity: 99.70% (data sheet), 99.97% (select batch)
- Appearance: Solid
- Color: White to off-white
- Solubility (in vitro): DMSO : 100 mg/mL (406.34 mM; need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.