Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3-Bromophenylacetic acid, 1878-67-7, MFCD00004330, 25g

Supplier:  Matrix Scientific 00323625G

Encompass_Preferred

Molecular Formula C8H7BrO2, Purity 98%, Molecular Weight 215.05, Melting Point 100-102°

Catalog No. 50-147-2258


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.