Learn More
Matrix Scientific 3-Bromophenylboronic acid, 89598-96-9, MFCD00239386, 5g
Supplier: Matrix Scientific 0074015G
Molecular Formula C6H6BBrO2, Purity 98%, Molecular Weight 200.83, Melting Point 164-168°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.