Learn More
Medchemexpress LLC 3-Bromophenylboronic acid | 89598-96-9 | 200.83 | 10 G

Supplier: Medchemexpress LLC HYW00471510G
3-Bromophenylboronic acid is a biochemical reagent used as a biological material or organic compound for life science research. It is a white to off-white solid, stable under recommended storage conditions, and soluble in DMSO.
- Purity of 99.89%
- Store powder at -20°C for up to 3 years or 4°C for up to 2 years
- Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
- Suitable for laboratory research applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.