Learn More
Medchemexpress LLC 3-Bromopropan-1-ol-d6 | 284474-43-7 | 145.03 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW007337S
Encompass_Preferred
3-Bromopropan-1-ol-d6 is a deuterium-labeled variant of 3-Bromopropan-1-ol, which is an endogenous metabolite. It is notably upregulated in buffaloes during the estrus period and is applicable in buffalo estrus research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-48356