Learn More
eMolecules​ 3-Bromopyrazolo[1,5-a]pyrimidin-7-amine | ChemScene | 1273577-17-5 | MFCD18633565 | 213.038 | C6H5BrN4 | 97.000 | Nc1ccnc2c(Br)cnn12 | 1g | 572196516
Supplier:  eMolecules​ 572196516
Encompass_Preferred
3-Bromopyrazolo[1,5-a]pyrimidin-7-amine | ChemScene | 1273577-17-5 | MFCD18633565 | 213.038 | C6H5BrN4 | 97.000 | Nc1ccnc2c(Br)cnn12 | 1g | 572196516
Catalog No. NC2223338