Learn More
Medchemexpress LLC 3-Bromopyridin-2-ol | 13466-43-8 | 174.00 | 250 G

Supplier: Medchemexpress LLC 250GHYW003045
3-Bromopyridin-2-ol is a biochemical reagent used as a biological material or organic compound for life science research and as a laboratory chemical.
- Purity of 99.6%
- Molecular weight of 174.00
- Chemical formula is C5H4BrNO
- Appears as a solid, white to light brown in color
- Melting point between 179-183°C
- Relative density of 1.776 g/cm3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.