Learn More
Medchemexpress LLC HY-19992 500mg , 3-Bromopyruvic acid CAS:1113-59-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19992500MG
Encompass_Preferred
HY-19992 500mg 3-Bromopyruvic acid CAS: 1113-59-3 3-Bromopyruvate (Bromopyruvic acid) is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. 3-Bromopyruvate inhibits cell growth and induces apoptosis through interfering with glycolysis. 3-Bromopyruvate induces autophagy by stimulating ROS formation in breast cancer cells. Antimicrobial activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1063