Learn More
Medchemexpress LLC 3-Bromothiophene | 872-31-1 | 99.8% | C4H3BrS | 10 G
SDP
Supplier:  Medchemexpress LLC HY5929910G
Encompass_Preferred
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive polymers with potential optoelectronic application value. It also serves as a key intermediate in the synthesis of various important pharmaceuticals, agrochemicals, and cosmetics. Additionally, 3-Bromothiophene can be used to synthesize chalcones and their derivatives, some of which demonstrate excellent anti-inflammatory, analgesic, and antibacterial activities.
  • Bromine atom-substituted 3-halogenated thiophene
  • Polymers are conductive with potential optoelectronic application value
  • Key intermediate in synthesis of pharmaceuticals, agrochemicals, and cosmetics
  • Used to synthesize chalcones and their derivatives
  • Derivatives demonstrate anti-inflammatory, analgesic, and antibacterial activities
Catalog No. 50-004-33984