Learn More
Medchemexpress LLC 3-Bromothiophene | 872-31-1 | 99.8% | C4H3BrS | 5 G
SDP
Supplier:  Medchemexpress LLC HY592995G
Encompass_Preferred
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive and have potential optoelectronic application value. It also serves as a key intermediate in the synthesis of various pharmaceuticals, agrochemicals, and cosmetics.
  • Used in the synthesis of chalcones and their derivatives
  • Derivatives exhibit anti-inflammatory, analgesic, and antibacterial activities
  • Polymers are conductive
  • Potential optoelectronic application value
  • Key intermediate for pharmaceuticals, agrochemicals, and cosmetics synthesis
Catalog No. 50-004-34042