Learn More
Medchemexpress LLC 3-Bromothiophene | 872-31-1 | 99.8% | C4H3BrS | 50 G
SDP
Supplier:  Medchemexpress LLC HY5929950G
Encompass_Preferred
3-Bromothiophene is a 3-halogenated thiophene with a bromine atom at the 3-position. Its polymers are conductive and show potential for optoelectronic applications. It also serves as a crucial intermediate in the creation of various pharmaceuticals, agrochemicals, and cosmetic products. This compound can be used in the synthesis of chalcones and their derivatives.
  • Polymers show potential for optoelectronic applications
  • Key intermediate for pharmaceuticals, agrochemicals, and cosmetics
  • Can be used in the synthesis of chalcones and their derivatives
  • Some derivatives show anti-inflammatory, analgesic, and antibacterial activities
Catalog No. 50-004-34402