Learn More
Medchemexpress LLC 3-buten-1-amine hydrochloride | 17875-18-2 | MFCD05663670 | >98.0% | 107.58 g/mol | C4H10ClN | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01932110G
Encompass_Preferred
3-Buten-1-amine hydrochloride is the hydrochloride salt of 3-buten-1-amine (but-3-en-1-amine). Supplied as a research chemical and synthetic intermediate, it is used in organic synthesis to prepare amine-containing building blocks, pharmaceuticals, and specialty chemicals.
  • High purity (>98.0%) suitable for laboratory research.
  • Primary amine functionality enables diverse coupling and derivatization reactions.
  • Stable hydrochloride salt for convenient handling and storage.
  • Available in small pack sizes for lab-scale work.
  • Molecular formula C4H10ClN and molecular weight 107.58 g/mol.
Catalog No. 50-002-99943