Learn More
Medchemexpress LLC 3-butenoic acid | 625-38-7 | MFCD00002782 | 98.6% | 86.09 g/mol | C4H6O2 | 1 G

Supplier: Medchemexpress LLC HYW0686971G
3-Butenoic acid (vinylacetic acid) is a small-molecule carboxylic acid used as a synthetic building block and intermediate in organic and medicinal chemistry research. It is supplied as a high-purity reagent for research and formulation applications.
- Small-molecule carboxylic acid used as a synthetic building block and intermediate.
- CAS number 625-38-7.
- Chemical formula C4H6O2; molecular weight 86.09 g/mol.
- Purity 98.6%.
- Soluble in DMSO at 100 mg/mL; suitable for in vitro formulation and certain in vivo formulations (e.g., ≥2.5 mg/mL in 10% DMSO/SBE-β-CD or 10% DMSO/corn oil).
- Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available in milligram to kilogram pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.