Learn More
Medchemexpress LLC 3-butenoic acid | 625-38-7 | MFCD00002782 | 98.6% | 86.09 g/mol | C4H6O2 | 100 G

Supplier: Medchemexpress LLC HYW068697100G
3-Butenoic acid (vinylacetic acid) is a small-molecule chemical intermediate used in organic synthesis and analytical applications. It is commonly employed as a building block for heterocycle formation and for derivatization in medicinal chemistry workflows.
- High purity: 98.6% suitable for research use.
- Molecular formula: C4H6O2, molecular weight: 86.09 g/mol.
- Identified by CAS number 625-38-7 for unambiguous substance identification.
- Available in multiple pack sizes including 100 G for preparative work.
- Supplied with supporting documents: data sheet, COA, SDS, and spectral files for quality verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.