Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-butenoic acid | 625-38-7 | MFCD00002782 | 98.6% | 86.09 g/mol | C4H6O2 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0686975G

Encompass_Preferred

3-butenoic acid (vinylacetic acid) is a low-molecular-weight unsaturated carboxylic acid used as a chemical intermediate and reagent in organic synthesis and research applications. It is typically supplied as a solid in laboratory pack sizes and is used for building block synthesis, medicinal chemistry, and other small-molecule research workflows.

  • Purity: 98.6%.
  • Cas number: 625-38-7.
  • Molecular formula: C4H6O2.
  • Molecular weight: 86.09 g/mol.
  • Common applications: organic synthesis and drug-intermediate chemistry.

Catalog No. 50-002-10257


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.