Learn More
Cayman Chemical 3-ButylIdenphthalIde 25g
SDP
Supplier:  Cayman Chemical 3498325 G
Encompass_Preferred
A plant metabolite with diverse biological activities; induces apoptosis and inhibits migration and EMT in 5637, BFTC-905, T24, and TCCSUP cells at 60 UG/ml; reduces tumor volume in a BFTC-905 mouse xenograft model at 100 and 200 mg/kg; increases life span and reduces spinal motor neuron loss in an SOD1G93A mouse model of ALS at 400 mg/kg; reduces polyQ accumulation and cerebral atrophy, as well as improves motor coordination in a mouse model of SCA3
Catalog No. 50-291-3129