Learn More
Medchemexpress LLC 3-carboxy-4-methyl-5-propyl-2-furanpropanoic acid | 86879-39-2 | 243.27 | C12H13D3O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6541191MG
Encompass_Preferred
CMPF-d3 is the deuterium-labeled isotopologue of 3-carboxy-4-methyl-5-propyl-2-furanpropanoic acid supplied as a research reagent for use as a tracer or internal standard in quantitative analytical methods. The d3 label provides a defined mass shift for reliable detection and quantitation in mass spectrometry workflows.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR.
  • Deuterium (d3) labeling produces a predictable mass shift for accurate quantitation.
  • Provided as a 1 mg solid for analytical method development and calibration.
  • Molecular formula C12H13D3O5; molecular weight 243.27.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-84837