Learn More
Medchemexpress LLC 3-chloro-4-methylphenyl boronic acid | 175883-63-3 | MFCD04039010 | 99.9% | 170.40 | C7H8BClO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01696310G
Encompass_Preferred
(3-Chloro-4-methylphenyl)boronic acid is an aryl boronic acid used as a building block in organic synthesis and life-science research. It is a solid with molecular formula C7H8BClO2, molecular weight 170.40 g/mol, and CAS 175883-63-3. Common applications include Suzuki-Miyaura cross-couplings and the preparation of functionalized aryl derivatives. Refer to the supplier datasheet for handling and storage instructions.
  • Aryl boronic acid reagent for organic synthesis.
  • Suitable for Suzuki-Miyaura cross-coupling reactions.
  • High chemical purity for reproducible results.
  • Solid form, easy to weigh and handle.
  • Stable at room temperature with defined storage guidance.
  • Provided with chemical identifiers for traceability.
Catalog No. 50-003-01231