Learn More
Cayman Chemical 3-chloro-L-TyrosIn 1g
SDP
Supplier:  Cayman Chemical 357531 G
Encompass_Preferred
A derivative of L-tyrosine; has been used as a biomarker of oxidative damage induced by MPO; plasma and atherosclerotic plaque levels are increased in patients with various cardiovascular diseases; plasma levels are increased in patients with colorectal cancer; formed in blood upon chlorine gas exposure and has been used as an indicator of chlorine poisoning during autopsy
Catalog No. 50-291-1209