Learn More
Medchemexpress LLC 3-chlorogentisyl alcohol | 32744-80-2 | 99.7% | 174.58 g/mol | C7H7ClO3 | 10 MG

Supplier: Medchemexpress LLC HY15116810MG
3-Chlorogentisyl alcohol is a small-molecule inhibitor of E. coli β-glucuronidase with reported antiproliferative activity. It is provided as a high-purity solid for biochemical and enzyme inhibition research.
- Potent E. coli β-glucuronidase inhibitor (IC50 0.74 μM; Ki 0.58 μM).
- Molecular formula C7H7ClO3; molecular weight 174.58 g/mol.
- Purity 99.7%.
- Available as solid quantities (including 10 mg) and as a 10 mM solution in DMSO.
- Appearance off-white to light yellow solid.
- Storage: powder -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.