Learn More
Medchemexpress LLC 3-Chlorogentisyl alcohol | 32744-80-2 | C7H7ClO3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151168
Encompass_Preferred
3-Chlorogentisyl alcohol is a potent E. coli β-glucuronidase inhibitor with an IC50 value of 0.74 μM and a Kᵢ value of 0.58 μM. It exhibits antiproliferative activity and holds potential for research in anti-cancer and anti-inflammatory therapies. The product is an off-white to light yellow solid.
  • Potent E. coli β-glucuronidase inhibitor
  • Exhibits antiproliferative activity
  • Potential for research in anti-cancer and anti-inflammatory therapies
  • Off-white to light yellow solid appearance
Catalog No. 50-003-74505