Learn More
Cayman Chemical 3-ChlorogentIsyl Alcohol 5mg
SDP
Supplier:  Cayman Chemical 406635 MG
Encompass_Preferred
A phenol and microbial metabolite with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 1 µM); acitve against MRSA (MIC = 50 UG/ml); active against 13 Gram-positive and Gram-negative bacteria at 50 UG/disc; has concentration-dependent antifouling activity against barnacle cyprids in the conventional submerged assay using polystyrene plates in a concentration-dependent manner; induces apoptosis and cell cycle arrest at the S phase in HeLa cancer cells at 50 µM; induces ssDNA breaks in a comet assay using HeLa cells at 35 µM
Catalog No. 50-294-5560