Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-chloropropane-1,2-diol dipalmitate | 51930-97-3 | MFCD09840071 | 98.3% | 587.36 g/mol | C35H67ClO4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW58581910MG

Encompass_Preferred

3-Chloropropane-1,2-diol dipalmitate is a fatty acid diester used as a tracer and analytical reagent in biochemical and lipid research. It is supplied as a white to off-white solid and is characterized by chemical formula C35H67ClO4 and molecular weight 587.36 g/mol. The material is intended for research use and requires low-temperature storage to preserve integrity.

  • Lipid diester reagent suitable as a tracer and analytical standard.
  • Chemical formula C35H67ClO4; molecular weight 587.36 g/mol.
  • Solid form, white to off-white, facilitating weighing and handling.
  • Store at -20°C under nitrogen; in solution store at -80°C up to 6 months or at -20°C up to 1 month.
  • Supplied in small research quantities for analytical and biochemical applications.

Catalog No. 50-002-84560


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.