Learn More
Medchemexpress LLC 3-cl-pyridine-amide-acrylaldehyde-piperazine | 99.3% | 294.74 | 25 MG
Supplier: Medchemexpress LLC 25MGHY176068
This product is a synthetic intermediate of LO-4-25 (HY-176067), an androgen receptor (AR) DNA binding domain ligand. LO-4-25 robustly degrades both AR and AR-V7 in 22Rv1 cells.
- Synthetic intermediate for LO-4-25.
- Androgen receptor (AR) DNA binding domain ligand precursor.
- Facilitates robust degradation of AR and AR-V7 in 22Rv1 cells.