Learn More
Medchemexpress LLC [3-Cyano-5-[[[2,4-dimethyl-5-[6-(3-pyridinyl)-1h-imidazo[1,2-b]pyrazol-1-yl]phenyl]amino]carbonyl]phen | 1448316-08-2 | 99.3% | 558.53 g/mol | C26H19F5N6OS | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10075625MG
Encompass_Preferred
BAY-826 is a small-molecule chemical probe that selectively and potently inhibits the receptor tyrosine kinase TIE-2 (Kd ≈ 1.6 nM). It is supplied for research use only and has been characterized in preclinical studies. The compound has molecular formula C26H19F5N6OS and a molecular weight of 558.53 g/mol.
  • Selective, potent TIE-2 inhibition with low nanomolar affinity.
  • Characterized for in vitro and in vivo preclinical studies.
  • Supplied as a solid and available in multiple pack sizes, with DMSO solutions for convenience.
  • High chemical purity suitable for research applications.
  • Molecular weight 558.53 g/mol and formula C26H19F5N6OS.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-13130