Learn More
Medchemexpress LLC 3-cyano-5-[[[2,4-dimethyl-5-[6-(3-pyridinyl)-1H-imidazo[1,2-b]pyrazol-1-yl]...phenyl]pentafluorosulfur | 1448316-08-2 | 99.3% | 558.53 g/mol | C26H19F5N6OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10075610MG
Encompass_Preferred
BAY-826 is a selective, potent small-molecule inhibitor of the TIE-2 (TEK) receptor tyrosine kinase used as a research chemical probe. It exhibits low-nanomolar binding (reported Kd ≈ 1.6 nM) and has been characterized in biochemical, cellular, and in vivo models. Supplied as a solid with high reported purity, it is intended for research use only.
  • Low-nanomolar potency against TIE-2 enabling sensitive biochemical assays.
  • High selectivity across kinases to reduce off-target activity.
  • Validated in cellular assays for target engagement.
  • Shown in vivo activity in preclinical models.
  • High reported purity suitable for pharmacology studies.
  • Soluble in DMSO for solution formulations.
Catalog No. 50-002-13158