Learn More
Medchemexpress LLC 3-Cyano-7-ethoxycoumarin | 117620-77-6 | 97.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD005510MM/1ML
Encompass_Preferred
3-Cyano-7-ethoxycoumarin is a fluorogenic cytochrome P-450 substrate that generates a blue fluorescent product upon enzyme cleavage. It is also a fluorescent probe useful in microsomal dealkylase studies and a high purity and quality chemical used as a molecular probe.
  • Can be used to study typical drug-drug interactions resulting from enzyme inhibition.
Catalog No. 50-004-10073