Learn More
Medchemexpress LLC 3-Cyano-7-ethoxycoumarin | 117620-77-6 | ≥98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD005550MG
Encompass_Preferred
3-Cyano-7-ethoxycoumarin is a fluorogenic cytochrome P-450 substrate that generates a blue fluorescent product upon enzyme cleavage. It is also a fluorescent probe useful in microsomal dealkylase studies and is a high purity and quality chemical used as a molecular probe. It can be used to study typical drug-drug interactions resulting from enzyme inhibition.
  • Fluorogenic cytochrome P-450 substrate
  • Generates blue fluorescent product upon enzyme cleavage
  • Fluorescent probe for microsomal dealkylase studies
  • High purity and quality chemical
  • Used as a molecular probe
  • Useful for studying drug-drug interactions from enzyme inhibition
Catalog No. 50-004-10320