Learn More
Medchemexpress LLC 3-cyano-L-phenylalanine | 57213-48-6 | MFCD01860885 | 99.8% | 190.20 g/mol | C10H10N2O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015177100G
Encompass_Preferred
H-Phe(3-CN)-OH (3-cyano-L-phenylalanine) is a phenylalanine derivative used as a research reagent in peptide synthesis, amino acid modification, and biochemical studies. The compound is provided as a high-purity solid suitable for analytical, synthetic, and biochemical applications.
Catalog No. 50-003-02270