Learn More
ILEX LIFE SCIENCES LLC 3-D LIFE PEG-LINK 200 UL
SDP
Supplier:  ILEX LIFE SCIENCES LLC L501
Encompass_Preferred
3-D Life PEG-Link, non-cell-degradable thiol-modified polyethylene glycol, hydrogel crosslinker component (200 ul). INTRO: 3-D Life technology, manufactured in Germany by Cellendes GmbH, is an integrated system for generation of hydrogels used in 3D cell culture. 3-D Life represents the most flexible system for biomimetic hydrogel design. DESCRIPTION: 3-D Life PEG-Link consists of polyethylene glycol carrying a thiol at each end. When combined with the 3-D Life Hydrogel polymers, thiol groups on PEG-Link form stable thioether bonds with the thiol-reactive groups on the polymers, resulting in formation of the hydrogel. PEG-Link can be used with 3-D Life functionalized polymers Mal-PVA, Mal-Dextran, SG-PVA and SG-Dextran. APPLICATION: 3-D Life PEG-Link is used together with thiol-reactive polymers of the 3-D Life system to establish 3-D cell cultures. QUANTITY: When used with polymers of the 3-D Life Hydrogel system up to 2 ml hydrogel can be generated, depending on gel stiffness.
Catalog No. 50-273-3915